3-Amino-5-fluor-4-methylbenzotrifluorid

Lieferant: COMBI-BLOCKS

AN-1351-100M
COBBAN-1351-100MEA 497 EUR
COBBAN-1351-100M
3-Amino-5-fluor-4-methylbenzotrifluorid
3-Amino-5-fluor-4-methylbenzotrifluorid
Formel: C₈H₇F₄N
MDL: MFCD11053832
CAS-Nummer: 1065073-89-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR