3-Amino-N-(3-chlorphenyl)benzolsulfonamid

Lieferant: COMBI-BLOCKS

AN-1368-5G
COBBAN-1368-5GEA 137 EUR
COBBAN-1368-5G
3-Amino-N-(3-chlorphenyl)benzolsulfonamid
3-Amino-N-(3-chlorphenyl)benzolsulfonamid
Formel: C₁₂H₁₁ClN₂O₂S
MDL: MFCD03147574
CAS-Nummer: 438017-93-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR