3-Amino-N-(4-fluorphenyl)benzolsulfonamid

Lieferant: COMBI-BLOCKS

AN-1377-5G
COBBAN-1377-5GEA 149 EUR
COBBAN-1377-5G
3-Amino-N-(4-fluorphenyl)benzolsulfonamid
3-Amino-N-(4-fluorphenyl)benzolsulfonamid
Formel: C₁₂H₁₁FN₂O₂S
MDL: MFCD09731646
CAS-Nummer: 953717-92-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR