3-Brom-5-chloranilin

Lieferant: COMBI-BLOCKS

Synonyme: 3-Bromo-5-chlorophenylamine

AN-1410-5G
COBBAN-1410-5GEA 88.6 EUR
COBBAN-1410-5G
3-Brom-5-chloranilin
3-Brom-5-chloranilin
Formel: C₆H₅BrClN
Molecular Weight: 206,47 g/mol
MDL: MFCD06797804
CAS-Nummer: 96558-78-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR