2-Nitro-4-(trifluormethoxy)anilin

Lieferant: COMBI-BLOCKS

AN-1445-5G AN-1445-25G AN-1445-100G
COBBAN-1445-5GEA 0 EUR
COBBAN-1445-5G COBBAN-1445-25G COBBAN-1445-100G
2-Nitro-4-(trifluormethoxy)anilin
2-Nitro-4-(trifluormethoxy)anilin
Formel: C₇H₅F₃N₂O₃
MDL: MFCD00042326
CAS-Nummer: 2267-23-4
UN: 2811
ADR: 6.1,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR