5-Brom-2-nitroanisol

Lieferant: COMBI-BLOCKS

Synonyme: 4-Bromo-2-methoxy-1-nitrobenzene

AN-1462-5G AN-1462-1G
COBBAN-1462-5GEA 359 EUR
COBBAN-1462-5G COBBAN-1462-1G
5-Brom-2-nitroanisol
5-Brom-2-nitroanisol
Formel: C₇H₆BrNO₃
Molecular Weight: 232.03 g/mol
MDL: MFCD09966092
CAS-Nummer: 103966-66-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR