N-Boc-5-brom-2,3-difluoranilin

Lieferant: COMBI-BLOCKS

Synonyme: tert-Butyl (5-bromo-2,3-difluorophenyl)carbamate

AN-1465-250M
COBBAN-1465-250MEA 623 EUR
COBBAN-1465-250M
N-Boc-5-brom-2,3-difluoranilin
N-Boc-5-brom-2,3-difluoranilin
Formel: C₁₁H₁₂BrF₂NO₂
Molecular Weight: 308,1193 g/mol
MDL: MFCD12025956
CAS-Nummer: 1150114-27-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR