2-Nitro-4-(trifluormethyl)anisol
Lieferant: COMBI-BLOCKS
Synonyme:
4-Methoxy-3-nitrobenzotrifluorid, 1-Methoxy-2-nitro-4-(trifluormethyl)benzol
AN-1476-5G
AN-1476-100G
AN-1476-25G
COBBAN-1476-5GEA
25.1
EUR
COBBAN-1476-5G
COBBAN-1476-100G
COBBAN-1476-25G
2-Nitro-4-(trifluormethyl)anisol
2-Nitro-4-(trifluormethyl)anisol
Formel:
C₈H₆F₃NO₃ Molecular Weight: 221.14 g/mol Siedepunkt: 90…95 °C (0.5 mmHg) Schmelzpunkt: 47 |
MDL:
MFCD00007099 CAS-Nummer: 394-25-2 EINECS: 206-891-4 UN: 1759 ADR: 8 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...