4'-Brom-2'-methylacetanilid

Lieferant: COMBI-BLOCKS

AN-1549-5G AN-1549-25G
COBBAN-1549-5GEA 0 EUR
COBBAN-1549-5G COBBAN-1549-25G
4'-Brom-2'-methylacetanilid
4'-Brom-2'-methylacetanilid
Formel: C₉H₁₀BrNO
MDL: MFCD00017779
CAS-Nummer: 24106-05-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR