4-Chlor-2-nitrophenol

Lieferant: COMBI-BLOCKS

AN-1560-5G
COBBAN-1560-5GEA 26.2 EUR
COBBAN-1560-5G
4-Chlor-2-nitrophenol
4-Chlor-2-nitrophenol
Formel: C₆H₄ClNO₃
Molecular Weight: 173,56 g/mol
Schmelzpunkt: 85…88 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00007113
CAS-Nummer: 89-64-5
EINECS: 201-927-5
UN: 2811
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR