N-Boc-4-brom-2-methylanilin

Lieferant: COMBI-BLOCKS

Synonyme: tert-Butyl (4-bromo-2-methylphenyl)carbamate, tert-Butyl 4-bromo-2-methylphenylcarbamate, 4-Bromo-2-methylaniline, N-BOC protected

AN-1569-5G AN-1569-1G
COBBAN-1569-5GEA 236 EUR
COBBAN-1569-5G COBBAN-1569-1G
N-Boc-4-brom-2-methylanilin
N-Boc-4-brom-2-methylanilin
Formel: C₁₂H₁₆BrNO₂
Molecular Weight: 286.1700 g/mol
MDL: MFCD01765429
CAS-Nummer: 306937-14-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR