3-Chlor-5-fluorphenol

Lieferant: COMBI-BLOCKS

AN-2568-5G AN-2568-25G AN-2568-1G
COBBAN-2568-5GEA 193 EUR
COBBAN-2568-5G COBBAN-2568-25G COBBAN-2568-1G
3-Chlor-5-fluorphenol
3-Chlor-5-fluorphenol
Formel: C₆H₄ClFO
Molecular Weight: 146.55 g/mol
MDL: MFCD00042212
CAS-Nummer: 202982-70-5
UN: 3265
ADR: 8 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR