5-Brom-4-methoxy-2-nitroanilin

Lieferant: COMBI-BLOCKS

AN-2571-5G AN-2571-1G AN-2571-250M
COBBAN-2571-5GEA 2040 EUR
COBBAN-2571-5G COBBAN-2571-1G COBBAN-2571-250M
5-Brom-4-methoxy-2-nitroanilin
5-Brom-4-methoxy-2-nitroanilin
Formel: C₇H₇BrN₂O₃
Molecular Weight: 247.0461 g/mol
MDL: MFCD00031127
CAS-Nummer: 6943-69-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR