5-Brom-4-methoxy-2-nitroanilin
Lieferant: COMBI-BLOCKS
AN-2571-5G
AN-2571-1G
AN-2571-250M
COBBAN-2571-5GEA
2040
EUR
COBBAN-2571-5G
COBBAN-2571-1G
COBBAN-2571-250M
5-Brom-4-methoxy-2-nitroanilin
5-Brom-4-methoxy-2-nitroanilin
Formel:
C₇H₇BrN₂O₃ Molecular Weight: 247.0461 g/mol |
MDL:
MFCD00031127 CAS-Nummer: 6943-69-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...