4-Brom-3,5-xylidin

Lieferant: COMBI-BLOCKS

Synonyme: 4-Bromo-3,5-dimethylaniline

AN-2600-1G
COBBAN-2600-1GEA 28.1 EUR
COBBAN-2600-1G
4-Brom-3,5-xylidin
4-Brom-3,5-xylidin
Formel: C₈H₁₀BrN
Molecular Weight: 200,0800 g/mol
MDL: MFCD07780648
CAS-Nummer: 59557-90-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR