5-Brom-2,4-dichloranilin

Lieferant: COMBI-BLOCKS

AN-2608-5G AN-2608-1G
COBBAN-2608-5GEA 0 EUR
COBBAN-2608-5G COBBAN-2608-1G
5-Brom-2,4-dichloranilin
5-Brom-2,4-dichloranilin
Formel: C₆H₄BrCl₂N
Molecular Weight: 240.91 g/mol
MDL: MFCD11226285
CAS-Nummer: 258344-01-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR