2-Brom-6-nitro-4-(trifluormethoxy)anilin

Lieferant: COMBI-BLOCKS

Synonyme: 2-Bromo-4-(trifluoromethoxy)-6-nitroaniline

AN-2635-25G
COBBAN-2635-25GEA 401 EUR
COBBAN-2635-25G
2-Brom-6-nitro-4-(trifluormethoxy)anilin
2-Brom-6-nitro-4-(trifluormethoxy)anilin
Formel: C₇H₄BrF₃N₂O₃
Molecular Weight: 301,0200 g/mol
MDL: MFCD04039212
CAS-Nummer: 886499-21-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR