4-Chlor-3,5-difluoranilin

Lieferant: COMBI-BLOCKS

AN-2664-1G
COBBAN-2664-1GEA 118 EUR
COBBAN-2664-1G
4-Chlor-3,5-difluoranilin
4-Chlor-3,5-difluoranilin
Formel: C₆H₄ClF₂N
Molecular Weight: 163,552 g/mol
MDL: MFCD09833604
CAS-Nummer: 2613-33-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR