5-Brom-2,3-difluoranilin

Lieferant: COMBI-BLOCKS

AN-2686-1G
COBBAN-2686-1GEA 774 EUR
COBBAN-2686-1G
5-Brom-2,3-difluoranilin
5-Brom-2,3-difluoranilin
Formel: C₆H₄BrF₂N
MDL: MFCD22205725
CAS-Nummer: 1375068-68-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR