2-Methyl-6-nitroanisol

Lieferant: COMBI-BLOCKS

AN-2890-1G
COBBAN-2890-1GEA 55.3 EUR
COBBAN-2890-1G
2-Methyl-6-nitroanisol
2-Methyl-6-nitroanisol
Formel: C₈H₉NO₃
MDL: MFCD00661626
CAS-Nummer: 18102-29-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR