Ethyl-3-amino-5-brombenzoat

Lieferant: COMBI-BLOCKS

AN-3030-1G
COBBAN-3030-1GEA 375 EUR
COBBAN-3030-1G
Ethyl-3-amino-5-brombenzoat
Ethyl-3-amino-5-brombenzoat
Formel: C₉H₁₀BrNO₂
MDL: MFCD12923267
CAS-Nummer: 690260-95-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR