3-Fluor-6-methyl-2-nitroanilin

Lieferant: COMBI-BLOCKS

AN-3229-1G
COBBAN-3229-1GEA 497 EUR
COBBAN-3229-1G
3-Fluor-6-methyl-2-nitroanilin
3-Fluor-6-methyl-2-nitroanilin
Formel: C₇H₇FN₂O₂
MDL: MFCD22383663
CAS-Nummer: 1393442-58-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR