1-(2-Methoxyethoxy)-4-nitrobenzol

Lieferant: COMBI-BLOCKS

AN-3238-25G
COBBAN-3238-25GEA 384 EUR
COBBAN-3238-25G
1-(2-Methoxyethoxy)-4-nitrobenzol
1-(2-Methoxyethoxy)-4-nitrobenzene
Formel: C₉H₁₁NO₄
Molecular Weight: 197,1879 g/mol
MDL: MFCD00463497
CAS-Nummer: 22483-40-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR