4-Methoxy-m-toluidin

Lieferant: COMBI-BLOCKS

Synonyme: 4-Methoxy-3-methylphenylamine, 4-Methoxy-3-methylaniline

AN-3691-5G
COBBAN-3691-5GEA 94.4 EUR
COBBAN-3691-5G
4-Methoxy-m-toluidin
4-Methoxy-m-toluidin
Formel: C₈H₁₁NO
Molecular Weight: 137,1800 g/mol
MDL: MFCD07779234
CAS-Nummer: 136-90-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR