1H-Indazol-5-boronsäure

Lieferant: COMBI-BLOCKS

BB-0106-1G
COBBBB-0106-1GEA 210 EUR
COBBBB-0106-1G
1H-Indazol-5-boronsäure
1H-Indazole-5-boronic acid
Formel: C₇H₇BN₂O₂
Molecular Weight: 161,9537 g/mol
MDL: MFCD06739151
CAS-Nummer: 338454-14-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR