1H-Indazol-6-boronsäure

Lieferant: COMBI-BLOCKS

BB-0147-1G
COBBBB-0147-1GEA 0 EUR
COBBBB-0147-1G
1H-Indazol-6-boronsäure
1H-Indazole-6-boronic acid
Formel: C₇H₇BN₂O₂
MDL: MFCD04972419
CAS-Nummer: 885068-10-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR