3-Amino-5-nitrophenylboronsäurepinakolester

Lieferant: COMBI-BLOCKS

BB-2004-1G
COBBBB-2004-1GEA 0 EUR
COBBBB-2004-1G
3-Amino-5-nitrophenylboronsäurepinakolester
3-Amino-5-nitrophenylboronsäurepinakolester
Formel: C₁₂H₁₇BN₂O₄
MDL: MFCD07363740
CAS-Nummer: 871329-51-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR