3-Carboxy-5-nitrophenylboronsäurepinakolester

Lieferant: COMBI-BLOCKS

BB-2005-5G BB-2005-1G
COBBBB-2005-5GEA 0 EUR
COBBBB-2005-5G COBBBB-2005-1G
3-Carboxy-5-nitrophenylboronsäurepinakolester
3-Carboxy-5-nitrophenylboronsäurepinakolester
Formel: C₁₃H₁₆BNO₆
MDL: MFCD03411932
CAS-Nummer: 377780-80-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR