3-Amino-5-nitrophenylboronsäure Hydrochlorid

Lieferant: COMBI-BLOCKS

Synonyme: 3-Amino-5-nitrobenzeneboronic acid hydrochloride

BB-2008-1G
COBBBB-2008-1GEA 497 EUR
COBBBB-2008-1G
3-Amino-5-nitrophenylboronsäure Hydrochlorid
3-Amino-5-nitrophenylboronsäure Hydrochlorid
Formel: C₆H₈BClN₂O₄
Molecular Weight: 218,40 g/mol
MDL: MFCD03411933
CAS-Nummer: 389621-79-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR