1-Boc-2-indolboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: N-(tert-Butoxycarbonyl)-1H-indole-2-boronic acid, 1H-Indole-2-boronic acid, N-BOC protected, N-Boc-indole-2-boronic Acid, 1-(tert-Butoxycarbonyl)indole-2-boronic acid, N-Boc-indole-2-boronic acid

BB-2009-5G BB-2009-25G BB-2009-1G
COBBBB-2009-5GEA 137 EUR
COBBBB-2009-5G COBBBB-2009-25G COBBBB-2009-1G
1-Boc-2-indolboronsäure
1-Boc-2-indolboronsäure
Formel: C₁₃H₁₆BNO₄
Molecular Weight: 261.08 g/mol
MDL: MFCD02093045
CAS-Nummer: 213318-44-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR