4-Pentylphenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: 4-n-Pentylbenzeneboronic acid, (4-Pentylphenyl)boronic acid

BB-2022-1G
COBBBB-2022-1GEA 25.9 EUR
COBBBB-2022-1G
4-Pentylphenylboronsäure
4-Pentylphenylboronsäure
Formel: C₁₁H₁₇BO₂
Molecular Weight: 192,07 g/mol
MDL: MFCD00995151
CAS-Nummer: 121219-12-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR