3-Chlor-4-methylphenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: 3-Chloro-4-methylphenylboronic Acid, 3-Chloro-4-methylbenzeneboronic acid

BB-2033-5G BB-2033-25G BB-2033-1G
COBBBB-2033-5GEA 97.9 EUR
COBBBB-2033-5G COBBBB-2033-25G COBBBB-2033-1G
3-Chlor-4-methylphenylboronsäure
3-Chlor-4-methylphenylboronsäure
Formel: C₇H₈BClO₂
Molecular Weight: 170.40 g/mol
MDL: MFCD04039010
CAS-Nummer: 175883-63-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR