4-Cyan-2-nitrophenylboronsäure
Lieferant: COMBI-BLOCKS
BB-2046-1G
BB-2046-5G
COBBBB-2046-1GEA
0
EUR
COBBBB-2046-1G
COBBBB-2046-5G
4-Cyan-2-nitrophenylboronsäure
4-Cyan-2-nitrophenylboronsäure
Formel:
C₇H₅BN₂O₄ |
MDL:
MFCD04115646 CAS-Nummer: 850568-46-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...