4-(2-Carboxyethyl)phenylboronsäure
Lieferant: COMBI-BLOCKS
Synonyme:
4-(2-Carboxyethyl)benzeneboronic acid
BB-2055-1G
BB-2055-5G
COBBBB-2055-1GEA
143
EUR
COBBBB-2055-1G
COBBBB-2055-5G
4-(2-Carboxyethyl)phenylboronsäure
4-(2-Carboxyethyl)phenylboronsäure
Formel:
C₉H₁₁BO₄ Molecular Weight: 193.99 g/mol |
MDL:
MFCD01318119 CAS-Nummer: 166316-48-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...