4,4'-Biphenyldiboronsäure
Lieferant: COMBI-BLOCKS
Synonyme:
[1,1'-Biphenyl]-4,4'-diyldiboronic acid
BB-2061-1G
COBBBB-2061-1GEA
63.4
EUR
COBBBB-2061-1G
4,4'-Biphenyldiboronsäure
4,4'-Biphenyldiboronsäure
Formel:
C₁₂H₁₂B₂O₄ Molecular Weight: 241,85 g/mol |
MDL:
MFCD00151795 CAS-Nummer: 4151-80-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...