Cyclopentylboronsäure

Lieferant: COMBI-BLOCKS

BB-2074-5G BB-2074-25G BB-2074-1G
COBBBB-2074-5GEA 119 EUR
COBBBB-2074-5G COBBBB-2074-25G COBBBB-2074-1G
Cyclopentylboronsäure
Cyclopentylboronic acid
Formel: C₅H₁₁BO₂
Molecular Weight: 113.95 g/mol
MDL: MFCD01074541
CAS-Nummer: 63076-51-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR