1-Boc-5-brom-2-indolboronsäure

Lieferant: COMBI-BLOCKS

BB-2086-5G BB-2086-25G BB-2086-1G
COBBBB-2086-5GEA 0 EUR
COBBBB-2086-5G COBBBB-2086-25G COBBBB-2086-1G
1-Boc-5-brom-2-indolboronsäure
1-Boc-5-brom-2-indolboronsäure
Formel: C₁₃H₁₅BBrNO₄
MDL: MFCD03701683
CAS-Nummer: 475102-13-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR