1-Boc-5-methoxy-2-indolboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: N-Boc-5-methoxy-2-indolylboronic acid, (1-(tert-Butoxycarbonyl)-5-methoxy-1H-indol-2-yl)boronic acid

BB-2087-5G BB-2087-1G BB-2087-25G
COBBBB-2087-5GEA 0 EUR
COBBBB-2087-5G COBBBB-2087-1G COBBBB-2087-25G
1-Boc-5-methoxy-2-indolboronsäure
1-Boc-5-methoxy-2-indolboronsäure
Formel: C₁₄H₁₈BNO₅
MDL: MFCD04039006
CAS-Nummer: 290331-71-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR