4-(Ethoxycarbonyl)phenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: 4-(Ethoxycarbonyl)benzeneboronic acid, (4-Ethoxycarbonylphenyl)boronic acid

BB-2113-5G BB-2113-25G BB-2113-1G
COBBBB-2113-5GEA 30.4 EUR
COBBBB-2113-5G COBBBB-2113-25G COBBBB-2113-1G
4-(Ethoxycarbonyl)phenylboronsäure
4-(Ethoxycarbonyl)phenylboronsäure
Formel: C₉H₁₁BO₄
Molecular Weight: 193.99 g/mol
MDL: MFCD02179441
CAS-Nummer: 4334-88-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR