3-(Methoxycarbonyl)phenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: Methyl 3-boronobenzoate, 3-(Methoxycarbonyl)benzeneboronic acid

BB-2116-5G BB-2116-25G BB-2116-1G BB-2116-100G
COBBBB-2116-5GEA 0 EUR
COBBBB-2116-5G COBBBB-2116-25G COBBBB-2116-1G COBBBB-2116-100G
3-(Methoxycarbonyl)phenylboronsäure
3-(Methoxycarbonyl)phenylboronsäure
Formel: C₈H₉BO₄
Molecular Weight: 179.97 g/mol
MDL: MFCD02093046
CAS-Nummer: 99769-19-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR