3-(Ethoxycarbonyl)phenylboronsäure
Lieferant: COMBI-BLOCKS
Synonyme:
3-(Ethoxycarbonyl)benzeneboronic acid, 3-Ethoxycarbonylphenylboronic acid
BB-2117-25G
BB-2117-1G
BB-2117-5G
BB-2117-100G
COBBBB-2117-25GEA
142
EUR
COBBBB-2117-25G
COBBBB-2117-1G
COBBBB-2117-5G
COBBBB-2117-100G
3-(Ethoxycarbonyl)phenylboronsäure
3-(Ethoxycarbonyl)phenylboronsäure
Formel:
C₉H₁₁BO₄ Molecular Weight: 193.99 g/mol |
MDL:
MFCD02179444 CAS-Nummer: 4334-87-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...