2-Nitrophenylboronsäurepinakolester

Lieferant: COMBI-BLOCKS

BB-2120-1G BB-2120-5G
COBBBB-2120-1GEA 0 EUR
COBBBB-2120-1G COBBBB-2120-5G
2-Nitrophenylboronsäurepinakolester
2-Nitrophenylboronsäurepinakolester
Formel: C₁₂H₁₆BNO₄
MDL: MFCD02179447
CAS-Nummer: 190788-59-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR