2-(Methoxycarbonyl)phenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: 2-Methoxycarbonylphenylboronic acid, 2-(Methoxycarbonyl)benzeneboronic acid

BB-2125-1G BB-2125-5G BB-2125-25G
COBBBB-2125-1GEA 0 EUR
COBBBB-2125-1G COBBBB-2125-5G COBBBB-2125-25G
2-(Methoxycarbonyl)phenylboronsäure
2-(Methoxycarbonyl)phenylboronsäure
Formel: C₈H₉BO₄
Molecular Weight: 179.97 g/mol
MDL: MFCD02179452
CAS-Nummer: 374538-03-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR