4-Fluor-3-methylphenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: 3-Methyl-4-fluorophenylboronic acid, 4-Fluoro-3-methylbenzeneboronic acid

BB-2127-25G BB-2127-100G BB-2127-5G
COBBBB-2127-25GEA 127 EUR
COBBBB-2127-25G COBBBB-2127-100G COBBBB-2127-5G
4-Fluor-3-methylphenylboronsäure
4-Fluor-3-methylphenylboronsäure
Formel: C₇H₈BFO₂
Molecular Weight: 153.95 g/mol
MDL: MFCD01863527
CAS-Nummer: 139911-27-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR