2-Aminophenylboronsäure
Lieferant: COMBI-BLOCKS
Synonyme:
2-Aminobenzolboronsäure, Dihydroxy(2-aminophenyl)boran, (2-Aminophenyl)metaborsäure
BB-2139-1G
BB-2139-5G
COBBBB-2139-1GEA
0
EUR
COBBBB-2139-1G
COBBBB-2139-5G
2-Aminophenylboronsäure
2-Aminophenylboronsäure
Formel:
C₆H₈BNO₂ Molecular Weight: 136.95 g/mol |
MDL:
MFCD01074645 CAS-Nummer: 5570-18-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...