3-Aminophenylboronsäure
Lieferant: COMBI-BLOCKS
Synonyme:
Dihydroxy(3-aminophenyl)boran, 3-Aminobenzolboronsäure, (3-Aminophenyl)metaborsäure
BB-2141-25G
BB-2141-5G
BB-2141-1G
BB-2141-100G
COBBBB-2141-25GEA
218
EUR
COBBBB-2141-25G
COBBBB-2141-5G
COBBBB-2141-1G
COBBBB-2141-100G
3-Aminophenylboronsäure
3-Aminophenylboronsäure
Formel:
C₆H₈BNO₂ Molecular Weight: 136.95 g/mol |
MDL:
MFCD00007755 CAS-Nummer: 30418-59-8 EINECS: 250-189-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...