3-Aminophenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: Dihydroxy(3-aminophenyl)boran, 3-Aminobenzolboronsäure, (3-Aminophenyl)metaborsäure

BB-2141-25G BB-2141-5G BB-2141-1G BB-2141-100G
COBBBB-2141-25GEA 218 EUR
COBBBB-2141-25G COBBBB-2141-5G COBBBB-2141-1G COBBBB-2141-100G
3-Aminophenylboronsäure
3-Aminophenylboronsäure
Formel: C₆H₈BNO₂
Molecular Weight: 136.95 g/mol
MDL: MFCD00007755
CAS-Nummer: 30418-59-8
EINECS: 250-189-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR