4-Methyl-2-nitrophenylboronsäure

Lieferant: COMBI-BLOCKS

BB-2147-1G
COBBBB-2147-1GEA 0 EUR
COBBBB-2147-1G
4-Methyl-2-nitrophenylboronsäure
4-Methyl-2-nitrophenylboronsäure
Formel: C₇H₈BNO₄
MDL: MFCD06659821
CAS-Nummer: 143697-03-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR