4-(Ethoxycarbonyl)-2-nitrophenylboronsäure

Lieferant: COMBI-BLOCKS

BB-2148-5G BB-2148-1G
COBBBB-2148-5GEA 0 EUR
COBBBB-2148-5G COBBBB-2148-1G
4-(Ethoxycarbonyl)-2-nitrophenylboronsäure
4-(Ethoxycarbonyl)-2-nitrophenylboronsäure
Formel: C₉H₁₀BNO₆
MDL: MFCD02179460
CAS-Nummer: 5785-70-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR