2-Hydroxyphenylboronsäure

Lieferant: COMBI-BLOCKS

BB-2159-5G BB-2159-1G BB-2159-25G BB-2159-100G
COBBBB-2159-5GEA 0 EUR
COBBBB-2159-5G COBBBB-2159-1G COBBBB-2159-25G COBBBB-2159-100G
2-Hydroxyphenylboronsäure
2-Hydroxyphenylboronsäure
Formel: C₆H₇BO₃
MDL: MFCD01074581
CAS-Nummer: 89466-08-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR